Order Entry
Puerto Rico
ContactUsLinkComponent
Silver nitrate 0.250 N (N/4) in aqueous solution
Irritant
Environment
Silver nitrate 0.250 N (N/4) in aqueous solution
Supplier:  Ricca Chemical
Certificates
undefined
Irritant
Environment
Silver nitrate 0.250 N (N/4) in aqueous solution
Supplier:  Ricca Chemical
Certificates

Specification Test Results

  • Appearance:
    Clear, colorless, odorless
  • Assay at 20 °C (traceable to NIST SRM 999):
    0.2500 ± 0.0001 N at 20.0 °C

Specification Test Results

Related Information