
Sodium hydroxide 0.100 N (N/10) in denatured alcohol
Catalog # RC7477-32
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly
- Size:1 L
- Cat. No.:RC7477-32
- Supplier no.:R7477000-1A
Specifications
Specification Test Results
- Appearance:Clear, colorless, alcohol odor
- Assay at 20 °C (traceable to NIST SRM 723):0.1000 ± 0.0005 N at 20.0 °C
Specification Test Results
About this item
:
:40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:2924
ADR:3,II
:GHS02
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548