Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium permanganate 0.100 N (N/10) in aqueous solution analytical reagent
Environment
Potassium permanganate 0.100 N (N/10) in aqueous solution analytical reagent
Supplier:  Ricca Chemical
Certificates
undefined
Environment
Potassium permanganate 0.100 N (N/10) in aqueous solution analytical reagent
Supplier:  Ricca Chemical
Certificates

Specification Test Results

  • Appearance:
    Clear, purple, odorless
  • Assay at 20 °C (traceable to NIST SRM 40):
    0.1000 ± 0.0001 N at 20.0 °C

Specification Test Results