
Sodium hydroxide 0.600 N in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Density:1.04 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:3266
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 84):0.6000 ± 0.0005 N at 20.0 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R7416000
SDS
VWR GHS Compliance Letter (US)