Sodium hydroxide 0.500 N (N/2) in methanol
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:3286
ADR:3,II
Specification Test Results
- Appearance:Clear, colorless, alcohol odor
- Assay at 20 °C (traceable to NIST SRM 723):0.5000 ± 0.0005 N at 20.0 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R7473000
SDS
VWR GHS Compliance Letter (US)