
Ammonia solution 5% (v/v) in aqueous solution (1 + 19)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
EINECS:215-647-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, colorless, ammonia odor
- Assay at 20 °C (traceable to NIST SRM 723):5.0 ± 0.1 % (v/v)