
Potassium permanganate 0.0100 N (N/100) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
:GHS09
Specification Test Results
- Appearance:Clear, purple, odorless
- Certified Concentration:0.0100 ± 0.0001 N at 20 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberRICCA Chemicals Ready-to-Use Titrants and Standards