Order Entry
Puerto Rico
ContactUsLinkComponent
 
Lugol's iodine solution concentrate Weigert Formulation
 
undefined
Lugol's iodine solution concentrate Weigert Formulation
 

 

  • Appearance:
    Dark brown liquid
  • Assay (vs. Sodium Thiosulfate/Starch - NIST SRM 136):
    0.389-0.399 N at 20°C

 

Frequently Bought Together