
Thymol blue 0.04% (w/v) in alcohol indicator
RC8421-16
Small Business Enterprise
:
- Pack type:Poly
- Size:16 oz.
- Cat. No.:RC8421-16
- Supplier no.:R8421000-500A
- Appearance:Clear, purplish-red, alcohol odor
- Suitability for Use:Red (pH 9.2)
:
:466.6 g/mol
:Ambient
:200-973-3
:1170
:3,II
:GHS02
:76-61-9
:C₂₇H₃₀O₅S
:MFCD00005869