Phenol red sodium salt 0.1% (w/v) alcoholic solution USP Test Solution (TS)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:34487-61-1
:
Formula:C₁₉H₁₃NaO₅S
MDL Number:MFCD00066901
:376.35 g/mol
Storage Temperature:Ambient
EINECS:252-057-8
UN:1170
ADR:3,III
Specification Test Results
- Appearance:Clear, orange, alcohol odor
- Suitability for Use:Yellow (pH 8.2)