Orthoboric acid 4% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
:61.83 g/mol
EINECS:233-139-2
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless liquid
- Assay (vs. Sodium Hydroxide/Phenolphthalein):1.94-2.06 % (w/v)