Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Calcium sulphate saturated aqueous solution USP Test Solution (TS) analytical reagent

Calcium sulphate saturated aqueous solution USP Test Solution (TS) analytical reagent

Calcium sulphate saturated aqueous solution USP Test Solution (TS) analytical reagent
Supplier:  Ricca Chemical
Certificates
Calcium sulphate saturated aqueous solution USP Test Solution (TS) analytical reagent
Calcium sulphate saturated aqueous solution USP Test Solution (TS) analytical reagent

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Calcium sulphate (anhydrous) anhydrite, Gypsum
  • CAS Number:7778-18-9
  • Formula:CaO₄S
  • MDL Number:MFCD00010912
  • Molecular Weight:136.14 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-900-3
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear, colorless, odorless
  • Calcium:
    Present
  • Sulfate:
    Present
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.