Ammonia solution 6.00 N analytical reagent
Supplier: Ricca Chemical
About this item
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Melting Pt:∼ 0°C
- Density:0.96
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Melting Pt:∼ 0°C
- Density:0.96
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Colorless liquid
- Assay (vs. Sulfuric Acid/Methyl Red):5.995-6.015 N at 20°C
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: R0645000-4AItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: R0645000-500A
- Supplier Number: R0645000-1A
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



