
Sodium hydroxide 1% (w/v) in aqueous solution (10 g/L)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
:GHS07
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 723):1.000 ± 0.004 % (w/v)
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R7243000
SDS
VWR GHS Compliance Letter (US)