Nitric acid 0.3% (v/v) in aqueous solution (3 + 997)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 84):3.00 ± 0.06 mL Nitric Acid per 997 mL of Water