Titanium ICP-MS Standard, 1 ml = 1 mg Ti (1,000 ppm Ti)(NH₄)2TiF₆ in H₂O / trace HF, Ricca Chemical Company
RCMSTI1KW-100
Small Business Enterprise
:
- Concentration:1000 µg/ml
- Matrix:H₂O/tr. HF
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCMSTI1KW-100
- Element:Titanium
- Supplier no.:RMSTI1KW-100N