Silicon ICP-MS Standard, 1 ml = 1 mg Si (1,000 ppm Si)(NH₄)₂SiF₆ in H₂O / tr HF, Ricca Chemical Company
RCMSSI1KW-100
Small Business Enterprise
:
- Concentration:1000 µg/ml
- Matrix:H₂O/tr. HF
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCMSSI1KW-100
- Element:Silicon
- Supplier no.:RMSSI1KW-100N