

Silver nitrate 5% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
EINECS:231-853-9
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 999):5.00 ± 0.05 % (w/v)