Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Potassium Standards
Potassium ICP-MS Standards, RICCA Chemical Company
Potassium ICP-MS Standards, RICCA Chemical Company
:
Ricca Chemical
Potassium ICP-MS Standards, RICCA Chemical Company
:
Ricca Chemical
8218993.PDF
ghs_RMSK10KN
VWR GHS Compliance Letter (US)