Calcium ICP-MS Standard, 1 ml = 1 mg Ca (1,000 ppm Ca)CaCO₃ in 3% HNO₃, Ricca Chemical Company
RCMSCA1KN-100
Small Business Enterprise
:
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCMSCA1KN-100
- Element:Calcium
- Supplier no.:RMSCA1KN-100N