Mercury ICP Standard, 1 mL = 10 mg Hg (10000 ppm Hg)Hg in 10% HNO₃, Ricca Chemical Company
Catalog # RCPHG10KN-500
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:10000 µg/ml
- Matrix:10% HCl
- Pack type:Poly
- Size:500 ml
- Cat. No.:RCPHG10KN-500
- Element:Mercury
- Supplier no.:RPHG10KN-500N