Order Entry
Puerto Rico
ContactUsLinkComponent
Praseodymium ICP Standard, 1 mL = 10 mg Pr (10,000 ppm Pr)Pr6O11 in 5% HNO3, Ricca Chemical Company
Praseodymium ICP Standard, 1 mL = 10 mg Pr (10,000 ppm Pr)Pr6O11 in 5% HNO3, Ricca Chemical Company
Catalog # RCPPR10KN-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Praseodymium ICP Standard, 1 mL = 10 mg Pr (10,000 ppm Pr)Pr6O11 in 5% HNO3, Ricca Chemical Company
Catalog # RCPPR10KN-100
Supplier:  Ricca Chemical
Supplier Number:  RPPR10KN-100N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    10000 µg/ml
  • Matrix:
    5% HNO₃
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. No.:
    RCPPR10KN-100
  • Element:
    Praseodymium
  • Supplier no.:
    RPPR10KN-100N

Specifications

About this item