Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
 
Trichloroacetic acid (TCA) 5% (w/v) in aqueous solution
 
undefined
 
 
 
Trichloroacetic acid (TCA) 5% (w/v) in aqueous solution
 

 

  • Appearance:
    Clear, colorless, slight Trichloroacetic odor
  • Assay at 20 °C (traceable to NIST SRM 84):
    5.00 ± 0.04 % (w/v)