Manganese ICP-MS Standard, 1 ml = 1 mg Mn (1000 ppm Mn)Mn(C₂H₃O₂)₂ in 3% HNO₃, RICCA Chemical Company
Catalog # RCMSMN1KN-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCMSMN1KN-100
- Element:Manganese
- Supplier no.:RMSMN1KN-100N