Order Entry
Puerto Rico
ContactUsLinkComponent
Magnesium ICP-MS Standard, 1 ml = 1 mg Mg (1000 ppm Mg) MgO in 3% HNO₃, Ricca Chemical Company
Magnesium ICP-MS Standard, 1 ml = 1 mg Mg (1000 ppm Mg) MgO in 3% HNO₃, Ricca Chemical Company
Catalog # RCMSMG1KN-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Magnesium ICP-MS Standard, 1 ml = 1 mg Mg (1000 ppm Mg) MgO in 3% HNO₃, Ricca Chemical Company
Catalog # RCMSMG1KN-100
Supplier:  Ricca Chemical
Supplier Number:  RMSMG1KN-100N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    3% HNO₃
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. No.:
    RCMSMG1KN-100
  • Element:
    Magnesium
  • Supplier no.:
    RMSMG1KN-100N

Specifications

About this item