Order Entry
Puerto Rico
ContactUsLinkComponent
 
Environment
Potassium permanganate 0.5000 N ± 0.0025 in aqueous solution
Catalog # RC6411-5
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Environment
Potassium permanganate 0.5000 N ± 0.0025 in aqueous solution
Catalog # RC6411-5
Supplier:  Ricca Chemical
Supplier Number:  R6411000-20F
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Cubitainer®
  • Size:
    20 L
  • Cat. No.:
    RC6411-5
  • Supplier no.:
    R6411000-20F

Specifications

Specification Test Results

  • Appearance:
    Clear, purple, odorless
  • Assay at 20 °C (traceable to NIST SRM 40):
    0.5000 ± 0.0025 N at 20.0 °C

Specification Test Results

About this item

 :
 :158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
 :GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364