


Potassium chromate 10% (w/v) in aqueous solution USP Test Solution (TS) indicator, for argentometric titration chloride-free
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7789-00-6
:
Formula:K₂CrO₄
MDL Number:MFCD00011368
:194.19 g/mol
EINECS:232-140-5
UN:3287
ADR:6.1,III
:GHS08
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear, yellow, odorless
- Refractive Index:10.0 ± 0.3 % (w/v)