Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Potassium hydroxide 30% (w/v) in aqueous solution analytical reagent

Potassium hydroxide 30% (w/v) in aqueous solution analytical reagent

Supplier:   Ricca Chemical
Potassium hydroxide 30% (w/v) in aqueous solution analytical reagent
Supplier:  Ricca Chemical
Certificates
Potassium hydroxide 30% (w/v) in aqueous solution analytical reagent
Potassium hydroxide 30% (w/v) in aqueous solution analytical reagent

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1814
  • ADR:8,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear, colorless, odorless
  • Assay at 20 °C (traceable to NIST SRM 723):
    30.00 ± 0.06 % (w/v)
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Poly
    1 gal.
    Log in for availability
    Log in for availability
    Each (1US Gal)
    Each (1US Gal) - $267.00
    $267.00
    Each (1US Gal)
       $267.00

    Supplier Number: R6153000-4A
  • Poly
    16 oz.
    Log in for availability
    Log in for availability
    Each (16oz)
    Each (16oz) - $81.00
    $81.00
    Each (16oz)
       $81.00

    Supplier Number: R6153000-500A
  • Poly
    32 oz.
    Log in for availability
    Log in for availability
    Each (32oz)
    Each (32oz) - $128.00
    $128.00
    Each (32oz)
       $128.00

    Supplier Number: R6153000-1A

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.