Iodine (iodine-iodide) 1.00 N in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
:
MDL Number:MFCD00011355
Storage Temperature:Ambient
EINECS:231-442-4
UN:3264
ADR:8,II
Specification Test Results
- Appearance:Clear, dark brown, Iodine odor
- Assay at 20 °C (traceable to NIST SRM 136):1.000 ± 0.001 N at 20.0 °C