Order Entry
Puerto Rico
ContactUsLinkComponent
Iodine (iodine-iodide) 0.200 N (N/5) in aqueous solution
Iodine (iodine-iodide) 0.200 N (N/5) in aqueous solution
Supplier:  Ricca Chemical
Certificates
undefined
Iodine (iodine-iodide) 0.200 N (N/5) in aqueous solution
Supplier:  Ricca Chemical
Certificates

Specification Test Results

  • Appearance:
    Clear, dark brown, Iodine odor
  • Assay at 20 °C (traceable to NIST SRM 136):
    0.2000 ± 0.0002 N at 20.0 °C

Specification Test Results

Related Information