Order Entry
Puerto Rico
ContactUsLinkComponent
Iodine (iodine-iodide) 0.0250 N (N/40) in aqueous solution
Iodine (iodine-iodide) 0.0250 N (N/40) in aqueous solution
Supplier:  Ricca Chemical
Certificates
undefined
Iodine (iodine-iodide) 0.0250 N (N/40) in aqueous solution
Supplier:  Ricca Chemical
Certificates

Specification Test Results

  • Appearance:
    Clear, brown, Iodine odor
  • Assay at 20 °C (traceable to NIST SRM 136):
    0.02500 ± 0.00002 N at 20.0 °C

Specification Test Results

Related Information