Iodine (iodine-iodide) 0.0200 N (N/50) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
:
Storage Temperature:Ambient
EINECS:231-442-4
UN:3264
ADR:8,II
Specification Test Results
- Appearance:Clear, brown, Iodine odor
- Assay at 20 °C (traceable to NIST SRM 136):0.02000 ± 0.00002 N at 20.0 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberRICCA Chemicals Ready-to-Use Titrants and Standards