Order Entry
Puerto Rico
ContactUsLinkComponent
EDTA disodium salt dihydrate ≥99.5%
EDTA disodium salt dihydrate ≥99.5%
Supplier:  IBI Scientific
Certificates
undefined
EDTA disodium salt dihydrate ≥99.5%
Supplier:  IBI Scientific

Specification Test Results

  • Purity:
    ≥99.5%
  • Calcium:
    ≤0.005%
  • Chloride:
    ≤0.001%
  • Phosphate:
    ≤0.001%
  • Sulfate:
    ≤0.01%
  • Heavy Metals:
    ≤0.001%
  • Dnase Assay:
    None Detected
  • Rnase Assay:
    None Detected
  • Protease Assay:
    None Detected

Specification Test Results

Related Information