Order Entry
Puerto Rico
ContactUsLinkComponent
Orthoboric acid ≥99.5%
Health
Orthoboric acid ≥99.5%
Catalog # IB70096
Supplier:  IBI Scientific
CAS Number:  
undefined
Health
Orthoboric acid ≥99.5%
Catalog # IB70096
Supplier:  IBI Scientific
Supplier Number:  IB70096
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    2.5 kg
  • Cat. No.:
    IB70096
  • Supplier no.:
    70096

Specifications

Specification Test Results

  • Purity:
    ≥99.5%
  • Calcium:
    ≤0.005%
  • Chloride:
    ≤0.001%
  • Phosphate:
    ≤0.001%
  • Sulfate:
    ≤0.01%
  • Heavy Metals:
    ≤0.001%
  • Dnase Assay:
    None Detected
  • Rnase Assay:
    None Detected
  • Protease Assay:
    None Detected

Specification Test Results

About this item

 :
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1.44 g/cm³ (25 °C)
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
 :GHS08
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337