Ampule Snapper, Disposable, WHEATON®, DWK Life Sciences
66009-125
Small Business Enterprise
- Description:Ampule Snapper
- Cat. No.:66009-125
- For:1 - 2 ml ampules
- Supplier no.:177105
Disposable ampule snapper protects the user against glass splinters and specially designed for 1 to 2 ml samples.
It prevents cross contamination between samples.