Corning® Advanced Polyethylene CentriStar™ Caps for Centrifuge Tubes
Supplier: Corning
Certificates
About this item
The CentriStar™ cap has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.
This leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions.
Specifications
- Catalog No:
- 89093-188
- 89093-192
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa