WHEATON® Black Phenolic Screw Cap, Linerless
Catalog # 26675-942
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Cap Size:22-350
- Description:Screw Caps for Vacule Vials
- Material:Black Phenolic
- Cat. No.:26675-942
- Style:Solid Top
- Liner material:Without Liner
- Supplier no.:239853
Specifications
About this item
Rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work.
- Aluminum foil faced pulp board liners can be used with organic solvents
- Good resistance to hydrocarbons, oils, ketones and alcohols
- Use low density polyethylene (LDPE) lined caps with distilled water, analytical standards and reagents
- PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product
- Bonded PTFE faced silicone liner ideal for storage of volatile solvents, sensitive compounds and corrosive chemicals
- Poly-Seal liner (LDPE) has unique cone liner design and offers superior torque retention and excellent sealing characteristics
Note: It is recommended that liner be tested prior to use for leak resistant applications.