White Light Illuminator
White Light Illuminator
Catalog # 470014-592
Supplier: PROMARK INTERNATIONAL, INC.
Supplier #: 750428
Jump to:
Product Details & Documents
Provides the Perfect Amount of Light for Photographing Gels
With its large surface area and even distribution of white light, the large light table is ideal for viewing or photographing DNA or PAGE electrophoresis gels. 110/240 VAC with a 10' cord. Viewing surface size: 9 1/2"L x 6"W. Overall size: 9"L x 12 1/2"W x 2 1/4"H.
Specifications
- Description:White Light Illuminator