ContactUsLinkComponent
Wheatstone Bridge
Wheatstone Bridge
Catalog # 470149-160
undefined
Wheatstone Bridge
Catalog # 470149-160
Supplier Number:  P50407A
Specifications
  • Description:
    Wheatstone Bridge
  • Dimensions:
    109×10 cm (wooden base)
  • Environmentally Preferable:
  • Cat. No.:
    470149-160
  • Supplier no.:
    P50407A
Specifications
About this item
The Wheatstone Bridge is complete with binding posts.
  • One Length of AWG 30 Nickel Silver Wire
  • Scale Graduated in mm
  • Slide Knife-Edge Contact
  • Binding Posts
The apparatus consists of one length of AWG 30 nickel silver wire mounted on a scale graduated in mm. The scale features a slide knife-edge contact.
For known and unknown resistance, two strips of heavy aluminum are mounted in each of the two arms of the bridge.