PYREX® West Condenser
470200-312
: Corning
- Description:Pyrex® West Condenser
- Length:345 mm
- Cat. No.:470200-312
- Jacket length:200 mm
- Supplier no.:2700-200
This borosilicate condenser is designed with a heavy walled outer jacket to provide long life. The lighter walled inner tube provides efficient heat transfer. To reduce breakage, tabulations are located on the same side.