ContactUsLinkComponent
Unknown Resistance Board
Unknown Resistance Board
Catalog # 470016-288
Supplier:  Wards
undefined
Unknown Resistance Board
Catalog # 470016-288
Supplier:  Wards
Supplier Number:  PH1161E/UN
Specifications
  • Description:
    Unknown Resistance Board
  • Cat. No.:
    470016-288
  • Supplier no.:
    PH1161E
Specifications
About this item
Use this set of nine unknown resistances to work with your Wheatstone bridges.
  • Connect in series or parallel
Each resistor has a unique resistance, but all are 1/4 watt metal film with 1% accuracy. Resistances start with 1.21 KΩ to protect against overloads and range up to 90.9 KΩ at no particular interval. They are mounted visibly underneath a printed circuit board with connections through binding posts on top; the resistors are numbered 1 to 9.