ContactUsLinkComponent
Tri-fold Field Shovel
Tri-fold Field Shovel
Catalog # 470013-336
undefined
Tri-fold Field Shovel
Catalog # 470013-336
Supplier Number:  381299
Specifications
  • Length:
    23"
  • Cat. No.:
    470013-336
  • Weight:
    36 oz.
  • Supplier no.:
    S031
Specifications
About this item
This compact and lightweight folding shovel can be used at a 90° angle for chopping or straight for shovelling, or it can be securely locked in three positions to provide the best angle for the job. The blade also has a serrated edge for cutting roots. Folded 10".
  • Aligned with GLOBE protocols