Tooth, Adult Slide
Catalog # 470182-816
Supplier: Avantor
Specifications
- Description:Tooth, Adult Slide
- Cat. No.:470182-816
- Stain:Hematoxylin and Eosin
- Supplier no.:WARD470182-816
- Orientation:Longitudinal section
Specifications
About this item
Observe tooth within jaw microscopically
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. In situ in jaw with surrounding tissues.