ContactUsLinkComponent
Thyroid Gland, Mammal Slide
Thyroid Gland, Mammal Slide
Catalog # 470182-792
Supplier:  Avantor
undefined
Thyroid Gland, Mammal Slide
Catalog # 470182-792
Supplier:  Avantor
Specifications
  • Description:
    Thyroid Gland, Mammal Slide
  • Environmentally Preferable:
  • Cat. No.:
    470182-792
  • Stain:
    Hematoxylin and Eosin
  • Supplier no.:
    WARD470182-792
  • Orientation:
    Section
Specifications
About this item
Structures and components of the thyroid
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Shows hormone stored in follicles, as well as simple cuboidal cells, resting on reticular tissue, lining the follicles.