The Mystery of Lyle and Louise: The Shoe Must Fit Footprint Analysis
Catalog # 470017-554
Supplier: Avantor
Specifications
- Description:The Mystery of Lyle and Louise: The Shoe Must Fit Footprint Analysis
- No. of lab groups:6
- Cat. No.:470017-554
- Supplier no.:VXH-LL-FTP
Specifications
About this item
Shoeprints Can Tell a Story
- Grade Level 10-College
- Links to a real world scenario
Using a print collected at the crime scene your students will be able to determine a critical information about the criminal. Students will also use computer software to analyze and compare 3-D shoe castings. Includes materials for footprint castings, along with software created to measure and compare three-dimensional models of the evidence shoe and the casting taken at the crime scene.
Web access to fully illustrated teacher's manual with technique background.