ContactUsLinkComponent
The Mystery of Lyle and Louise: The Jagged Edge Glass Fragment Identification
The Mystery of Lyle and Louise: The Jagged Edge Glass Fragment Identification
Catalog # 470144-946
Supplier:  Avantor
undefined
The Mystery of Lyle and Louise: The Jagged Edge Glass Fragment Identification
Catalog # 470144-946
Supplier:  Avantor
Specifications
  • Description:
    The Mystery of Lyle and Louise: The Jagged Edge Glass Fragment Identification
  • No. of lab groups:
    6
  • Cat. No.:
    470144-946
  • Supplier no.:
    VXH-LL-GFI
Specifications
About this item
Analyze Refractive Index of Glass
  • Grade Level 10-College
  • Links to a real world scenario
When investigators discovered three deceased victims at a vehicle crash site over the side of Backbone Mountain, they retraced the vehicle's path and also discovered several pieces of broken glass lying in the roadway. By examining the refractive index of various glass fragments, students will learn the importance of glass analysis in the forensic field and how this analysis is performed by forensic scientists. In addition, students will learn to microscopically determine the refractive indices of several known types of glass, and then apply what they have learned in the comparison of glass fragments collected from a crime scene to a potential source.
Web access to fully illustrated teacher's manual with technique background.