Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:S. aureus: Freeze dried with media pack
- Storage conditions:Freezer
- Cat. No.:470179-564
- Packaging:Lyophilized
- Media:Tryptic soy agar
- Supplier no.:WARD470179-564
This yellow, shiny organism is known to cause wound infections and more famously, toxic shock syndrome.
- Biosafety Level: 2
- Canada: Containment Level 2 - PHAC Licence required
- Gram Stain: Positive
- Morphology: cocci in grape like clusters
- Coagulase: Positive
- Catalase: Positive
- Motility: Negative
- Respiration: Aerobic
- Chromogenesis: Yes (yellow-to-orange)
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.
Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Canada: Containment Level 2 – PHAC License required.
Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals.
Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.
As with all organisms, Ward's recommends that you utilize proper aseptic technique when handling all microorganisms.