Spermatic Cord Slide
Catalog # 470183-088
Supplier: Avantor
Specifications
- Description:Spermatic Cord
- Cat. No.:470183-088
- Stain:Hematoxylin and triosin
- Supplier no.:WARD470183-088
- Orientation:Section
Specifications
About this item
Male Reproductive System
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Ductus deferens, epididymal and testicular nerve, associated blood vessels, and connective tissues.