Specific Gravity Bottles
470120-780
Small Business Enterprise
- Capacity:25 ml
- Description:Specific gravity bottles
- Environmentally Preferable:
- Cat. No.:470120-780
- Supplier no.:3505-02
Also called density bottles or R.D. bottles, each has a perforated stopper (about 1 mm bore) and a ground mouth for proper fit. Made of Borosilicate glass. Uncalibrated.