Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Somso® Brain Stem Model

Brain Stem, 49, 3, 14 cm, 16 cm, 16 cm
Catalog # 470029-734
Supplier:   MARCUS SOMMER
 :  BS 23/2
Somso® Brain Stem Model
Catalog # 470029-734
Supplier:  MARCUS SOMMER
Somso® Brain Stem Model
Somso® Brain Stem Model

Brain Stem, 49, 3, 14 cm, 16 cm, 16 cm

Price
$528.00
Log in for availability
Each
Product Details & Documents

Illustrates Emotional And Involuntary Response Center Of The Brain

  • Removes From Base For Easy Study
  • Large Size Allows For Easy Differentiation Of Structures
  • 5 Year Manufacturers Warranty

Ideal for advanced anatomical study, the model focuses on the intricate structural details of the brain stem, sagitally sectioned, to reveal the pons, diencephalon, medulla, and cranial nerves. The two-piece model is mounted on a stand.

Specifications
  • Description:
    Brain Stem
  • Height:
    16 cm
  • Length:
    16 cm
  • No. of keyed items:
    49
  • Width:
    14 cm
  • No. of parts:
    3
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.